Learn More
Sodium tetrafluoroborate, 95%, Thermo Scientific Chemicals
Description
Fluorinating agentSodium tetrafluoroborate is used in fluxes, metal finishing and in plating circuits. It is used as a starting material in the preparation of ionic liquids as well as boron trifluoride. It acts as an efficient catalyst for the synthesis of 3,4-dihydropyrimidin-2(1H)-ones and thiones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 384°C (decomposition) |
Color | White |
Physical Form | Granular/Lump |
Quantity | 500 g |
Linear Formula | NaBF4 |
Merck Index | 14,8617 |
Solubility Information | Soluble in water. |
Formula Weight | 109.79 |
Percent Purity | 95% |
Sensitivity | Hygroscopic |
Show More |
Safety and Handling
Hazard Category | H314 |
Hazard Statement | GHS H Statement H318-H335-H315 Causes serious eye damage. May cause respiratory irritation. Causes skin irritation. |
Precautionary Statement | P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
DOTInformation | Hazard Class: 8; Packaging Group: II |
EINECSNumber | 237-340-6 |
RTECSNumber | ED2975000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.