Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate, 97%, may cont. 1-5% water

Catalog No. aah3001403
Click to view available options
Quantity:
1 g
5 g
25 g

Sodium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate, 97%, C32H12BF24Na, CAS Number-79060-88-1 | CAS: 79060-88-1 | C32H12BF24Na | 886.209 g/mol

Sodium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate is a lipophilic additive of tetraphenylborate and it is used for solvent extraction of cations and solvent polymeric membrane electrodes and optodes. It is also used as an anionic phase-transfer catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate is a lipophilic additive of tetraphenylborate and it is used for solvent extraction of cations and solvent polymeric membrane electrodes and optodes. It is also used as an anionic phase-transfer catalyst.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79060-88-1
Molecular Formula C32H12BF24Na
Molecular Weight (g/mol) 886.209
MDL Number MFCD00043323
InChI Key LTGMONZOZHXAHO-UHFFFAOYSA-N
Synonym sodium tetrakis 3,5-bis trifluoromethyl phenyl borate, nabarf, sodium tetrakis 3,5-bis trifluoromethyl phenyl boranuide, sodium2-sulfonatoethylmethacrylate, sodium tetrakis 3,5-bis trifluoromethyl phenyl bor, sodium tetrakis 3,5-bis-trifluoromethylphenyl borate, sodiotetrakis 3,5-bis trifluoromethyl phenyl boron v, sodium tetrakis 3,5-bis trifluoromethyl phenyl-borate, sodium tetrakis 3,5-bis trifluoromethyl phenyl borate 1-
PubChem CID 23681909
IUPAC Name sodium;tetrakis[3,5-bis(trifluoromethyl)phenyl]boranuide
SMILES [B-](C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F)(C2=CC(=CC(=C2)C(F)(F)F)C(F)(F)F)(C3=CC(=CC(=C3)C(F)(F)F)C(F)(F)F)C4=CC(=CC(=C4)C(F)(F)F)C(F)(F)F.[Na+]

Specifications

Melting Point 310°C (decomposition)
Quantity 1 g
Solubility Information Soluble in water.
Formula Weight 886.2
Percent Purity 97%
Assay may contain 1 to 5% water
Chemical Name or Material Sodium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.