Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium tetraphenylborate, ACS, 99.5% min

Catalog No. AA3643018
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3643018 50 g
AA3643004 2 g
AA3643009 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3643018 Supplier Thermo Scientific Chemicals Supplier No. 03643018
Only null left
Add to Cart

CAS: 143-66-8 | C24H20BNa | 342.22 g/mol

Sodium tetraphenylborate is used as a precipitating reagent for potassium, as a protecting agent to protect amino acids. It is used in the preparation of N-acylammonium salts. It finds application as a phenyl donor in palladium-catalyzed cross-coupling reactions to prepare arylalkenes and biaryl compounds. It is used in inorganic and organometallic chemistry due to its solubility in nonpolar solvents. Furthermore, it is involved in the isolation of complexes having dinitrogen ligands.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143-66-8
Molecular Formula C24H20BNa
Molecular Weight (g/mol) 342.22
MDL Number MFCD00011494
InChI Key HFSRCEJMTLMDLI-UHFFFAOYSA-N
Synonym sodium tetraphenylborate, tetraphenylboron sodium, sodium tetraphenylboron, kalignost, sodium tetraphenylborate 1-, kalibor, dotite kalibor, kariporu k, borate 1-, tetraphenyl-, sodium, sodium tetraphenylboride
PubChem CID 2723787
IUPAC Name sodium;tetraphenylboranuide
SMILES [Na+].C1=CC=C(C=C1)[B-](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point >300°C
Color White
pH 8
Linear Formula NaB(C6H5)4
Quantity 50 g
UN Number UN2811
Beilstein 3599783
Sensitivity Light sensitive
Merck Index 14,8690
Solubility Information Soluble in water and ethanol.
Formula Weight 342.23
Percent Purity ≤99.5%
Grade ACS
Physical Form Powder
Chemical Name or Material Sodium tetraphenylborate
Show More Show Less
Hazard Category H301-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 205-605-5
RTECSNumber ED3362500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.