Learn More
Sodium thiocyanate, 96%
Description
A compound used to convert alkyl halides into the corresponding alkylthiocyanates. Undergoes nucleophilic substitution reactions with phase-transfer catalysts. Silver thiocyanate may be used as well, the precipitation of insoluble silver halides help simplify workup. Treatment of isopropyl bromide with sodium thiocyanate in a hot ethanolic solution affords isopropyl thiocyanate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
A compound used to convert alkyl halides into the corresponding alkylthiocyanates. Undergoes nucleophilic substitution reactions with phase-transfer catalysts. Silver thiocyanate may be used as well, the precipitation of insoluble silver halides help simplify workup. Treatment of isopropyl bromide with sodium thiocyanate in a hot ethanolic solution affords isopropyl thiocyanate.
Solubility
Soluble in water (1250g/L), alcohol, and acetone.
Notes
Hygroscopic, protect from humidity and water. Store in cool, dry conditions in well sealed container.

Specifications
Specifications
Melting Point | 287°C |
Density | 1.74 |
pH | 6 to 8 |
Odor | Odorless |
Quantity | 1000 g |
Beilstein | 3594965 |
Sensitivity | Hygroscopic; Light sensitive |
Merck Index | 14,9327 |
Solubility Information | Soluble in water (1250g/L),alcohol,and acetone. |
Formula Weight | 81.07 |
Show More |
Safety and Handling
Hazard Category | H302+H312+H332-H373 |
Hazard Statement | GHS H Statement H302-H312-H332 Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled. |
Precautionary Statement | P260-P264b-P270-P271-P280-P302+P352-P304+P340-P314-P330-P362-P501c |
EINECSNumber | 208-754-4 |
RTECSNumber | XL2275000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.