Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium thiocyanate, 98+%, for analysis

Catalog No. AC446290010
Click to view available options
Quantity:
250 g
1 kg

CAS: 540-72-7 | CNNaS | 81.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 540-72-7
Molecular Formula CNNaS
Molecular Weight (g/mol) 81.07
MDL Number MFCD00011123
InChI Key VGTPCRGMBIAPIM-UHFFFAOYSA-M
Synonym sodium thiocyanate, sodium rhodanate, sodium sulfocyanate, sodium rhodanide, thiocyanic acid, sodium salt, thiocyanate sodium, scyan, sodium isothiocyanate, nascn, haimased
PubChem CID 516871
ChEBI CHEBI:30952
IUPAC Name sodium;thiocyanate
SMILES [Na+].[S-]C#N

Specifications

Melting Point 287.0°C
Quantity 1 kg
Assay Percent Range 98% min. (Argentometry)
Linear Formula NaSCN
Merck Index 15,948
Solubility Information Solubility in water: 1390g/L (21°C). Other solubilities: soluble in alcohol, freely soluble in acetone
Formula Weight 81.07
Percent Purity ≥98%
Grade Analytical
Packaging Glass bottle
Chemical Name or Material Sodium thiocyanate
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 208-754-4

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.