Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium thiocyanate, ACS, 98.0% min

Catalog No. AA3338822
Click to view available options
Quantity:
100 g
500 g
2 kg

CAS: 540-72-7 | CNNaS | 81.07 g/mol

Sodium thiocyanate is used as a precursor for the synthesis of pharmaceuticals and specialty chemicals. It is used to prepare alkylthiocyanates by reacting with alkyl halides. It is involved nucleophilic substitution reactions with phase-transfer catalysts. It is a common laboratory reagent used to test presence of Fe3+ ions. It acts as a spinning solvent for polyacrylonitrile in fiber industry, photographic stabilizer, corrosion inhibitor, polymer stabilizer and hardening accelerator for cement and concrete mixes. Further, it is also used in both the crystallization and purification of proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 540-72-7
Molecular Formula CNNaS
Molecular Weight (g/mol) 81.07
MDL Number MFCD00011123
InChI Key VGTPCRGMBIAPIM-UHFFFAOYSA-M
Synonym sodium thiocyanate, sodium rhodanate, sodium sulfocyanate, sodium rhodanide, thiocyanic acid, sodium salt, thiocyanate sodium, scyan, sodium isothiocyanate, nascn, haimased
PubChem CID 516871
ChEBI CHEBI:30952
SMILES [Na+].[S-]C#N

Specifications

Melting Point 287°C
Color White
Physical Form Crystals and/or Chunks
Quantity 100 g
Assay Percent Range 98% min.
Beilstein 3594965
Solubility Information Soluble in water,acetone,alcohol.Soluble in water,acetone,alcohols and ammonia.
IUPAC Name sodium;thiocyanate
Formula Weight 81.07
Grade ACS
Sensitivity Hygroscopic; Light sensitive
Appearance White crystalline/chunks
Density 1.74 g/cm3 at 20°C
Chemical Name or Material Sodium thiocyanate, For analysis ACS
Show More Show Less
Hazard Category H302+H312+H332-H373
Precautionary Statement P260-P264b-P270-P271-P280-P302+P352-P304+P340-P314-P330-P362-P501c
EINECSNumber 208-754-4
RTECSNumber XL2275000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.