Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Thiosulfate, 0.010 Normal (N/100), Reagents

Catalog No. 01334063
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
10 L
20 L
Packaging:
Cubitainer
Glass Bottle
Poly Bottle
Safety Coated Glass
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01-334-063 500 mL Poly Bottle
01-334-069 10 L Cubitainer
01-334-068 1 L Glass Bottle
01-334-067 2.5 L Safety Coated Glass
01-334-066 20 L Cubitainer
01-334-065 4 L Poly Bottle
01-334-064 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 01-334-063 Supplier Reagents Holdings Llc Supplier No. CS000100500A
Only null left
Add to Cart
Add to Cart

Sodium Thiosulfate, 0.010 Normal (N/100), Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.

Chemical Identifiers

CAS 7732-18-5 , 10102-17-7 , 497-19-8
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Sodium hyposulfite, Hypo, Hyposulfite of soda
IUPAC Name water
SMILES O

Specifications

Boiling Point 100°C
Color Colorless
Physical Form Liquid
Quantity 500 mL
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.001
Chemical Name or Material Sodium Thiosulfate, 0.010 Normal (N/100)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.