Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Thiosulfate, 0.100 Normal Standardized, NIST Traceable, ISO 17025, Reagents
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
10 L
20 L
Packaging:
Cubitainer
Glass Bottle
Poly Bottle
Specifications
Specifications
| Chemical Name or Material | Sodium Thiosulfate, 0.100 Normal Standardized, NIST Traceable, ISO 17025 |
| Boiling Point | 100°C |
| CAS | 7732-18-5, 10102-17-7, 10102-17-7 |
| Color | Colorless |
| Physical Form | Liquid |
| Quantity | 10 L |
| Molecular Formula | Na2S2O3/H2O |
| Synonym | Sodium hyposulfite, Hypo, Hyposulfite of soda |
| Grade | Reagent |
| Packaging | Cubitainer |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction