Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Thiosulfate 0.1N, LaMotte™
SDPSCI_ED

Catalog No. s72420 Shop All LaMotte Company Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S72420 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S72420 Supplier LaMotte Company Supplier No. 6155L
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7732-18-5
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide

Specifications

Boiling Point >100°C
Color Colorless
pH 7.0 to 7.5
Physical Form Liquid or Solid
Quantity 500 mL
Percent Purity 95 to 99%
Concentration or Composition (by Analyte or Components) 0.1N
Packaging Bottle
Chemical Name or Material Sodium Thiosulfate
Recommended Storage Keep containers tightly closed in a dry, cool and well-ventilated place. Store at room
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.