Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Thiosulfate, 10% (w/v), Ricca Chemical
SDP

Catalog No. p-3340423
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Natural Poly Bottle

Chemical Identifiers

CAS 10102-17-7, 7732-18-5
Molecular Formula H10Na2O8S2
Molecular Weight (g/mol) 248.172
InChI Key PODWXQQNRWNDGD-UHFFFAOYSA-L
Synonym sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate
PubChem CID 61475
ChEBI CHEBI:32150
IUPAC Name disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate
SMILES O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+]

Specifications

Color Colorless
CAS Min % 89.0, 8.91
CAS Max % 93.0, 9.28
Percent Purity 10%
Grade Laboratory
Concentration or Composition (by Analyte or Components) 10% (w/v) Aqueous Solution
Packaging Natural Poly Bottle
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Sodium Thiosulfate
ShelfLife 18 months
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.