Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium thiosulfate, 98.5%, extra pure, anhydrous, Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
500 g
1 kg
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
Specifications
Melting Point | 48.0°C |
Color | White |
pH | 6.0 to 8.5 (5% soln. at 20°C) |
Physical Form | Crystalline Powder |
Quantity | 500 g |
Assay Percent Range | 98% min. (on dry substance) (Iodimetry) |
Linear Formula | Na2S2O3 |
Merck Index | 15,8821 |
Solubility Information | Solubility in water: 700g/L (20°C). Other solubilities: practically insoluble in alcohol |
IUPAC Name | disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane |
Show More |
Safety and Handling
EINECSNumber | 231-867-5 |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
missing translation for 'productTitle'
Spot an opportunity for improvement?
missing translation for 'provideContentCorrection'