Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethylzinc, 0.9M (15 wt%) solution in hexane, AcroSeal™, Thermo Scientific Chemicals

Catalog No. AC205518000
Click to view available options
Quantity:
15 g
100 g
100 mL
800 mL
Packaging:
AcroSeal glass bottle
AcroSeal™ Glass Bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diethylzinc
Color Brown-Gray
Name Note 1M Solution in Hexane
CAS 557-20-0, 64742-49-0
Boiling Point 118.0°C
Melting Point -39.0°C to +28.0°C
Molecular Formula C4H10Zn
Linear Formula (C2H5)2Zn
MDL Number MFCD00009021
Concentration 1M, exact strength on the certificate of analysis
Merck Index 15, 3151
Density 0.7260g/mL
Synonym diethylzinc, zinc, diethyl, diethyl zinc, zinc ethide, zinc ethyl, unii-s0w5nqh7c6, et2zn, znet2, s0w5nqh7c6, c2h5 2zn
Solubility Information Solubility in water: reacts.
InChI Key HQWPLXHWEZZGKY-UHFFFAOYSA-N
SMILES CC[Zn]CC
IUPAC Name diethylzinc
Molecular Weight (g/mol) 123.50
PubChem CID 11185
Formula Weight 123.5
Packaging AcroSeal™ Glass Bottle
Specific Gravity 0.726
Viscosity 0.7 mPa.s (20°C)
Quantity 800 mL
Flash Point −40°C
Physical Form Solution
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.