Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisher Science Education™ Sodium Thiosulfate, Pentahydrate
Shop All Fisherbrand Products

Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
Packaging:
Reagent Grade
Description
This product is for educational use - no C of A is available.
Specifications
Specifications
Color | White |
Quantity | 500 g |
Assay Percent Range | >95% |
Linear Formula | Na2S2O3·5H2O |
Formula Weight | 248.18 |
Packaging | Reagent Grade |
Chemical Name or Material | Sodium Thiosulfate, Pentahydrate |
Safety and Handling

ShelfLife | 1 year |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction