Learn More
Sodium thiosulfate pentahydrate, ACS, 99.5-101.0%
Description
Sodium thiosulfate pentahydrate is used as an analytical reagent in laboratories. It is used in photographic processing and in gold extraction. Its alkylated derivatives, S-alkylthiosulfonates, are known as Bunte salts. It is involved in the preparation of thioglycolic acid. Furthermore, it is used to dechlorinate tap water, for removing free bromine from solutions and to check the pH of bleach solutions with liquid indicators. Other uses include tanning of leather; as an active component in hand warmers and chemical heating pads and as an anionic surfactant to aid in the dispersion of pharmaceutical preparations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 100°C-5H{2}O |
| Melting Point | 48°C |
| Color | Colorless |
| Physical Form | Crystalline |
| Quantity | 500 g |
| Assay Percent Range | 99.5-101.0% |
| Solubility Information | Soluble in water and ammonia. Insoluble in alcohol and organic solvents. |
| Formula Weight | 248.17 (158.10 Anhydrous) |
| Grade | ACS |
| Odor | Odorless |
| Show More |
Safety and Handling
| Hazard Category | H500 |
| EINECSNumber | 231-867-5 |
| RTECSNumber | WE6660000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.