Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium triacetoxyborohydride, 95%

Catalog No. p-7026197
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 56553-60-7 | C6H10BNaO6 | 211.94 g/mol

Sodium triacetoxyborohydride is used as a reagent in reductive amination of ketones, aldehydes and lactamization of carbonyl compounds with amines. It also serves as a reducing agent in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium triacetoxyborohydride is used as a reagent in reductive amination of ketones, aldehydes and lactamization of carbonyl compounds with amines. It also serves as a reducing agent in organic synthesis.

Solubility
Soluble in dimethyl sulfoxide, methanol, benzene, toluene, terahydrofuran, dioxane and methylene chloride.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents, water, alcohols and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56553-60-7
Molecular Formula C6H10BNaO6
Molecular Weight (g/mol) 211.94
MDL Number MFCD00012211
InChI Key HHYFEYBWNZJVFQ-UHFFFAOYSA-N
Synonym c6h10bnao6, sodium triacetoxyboron 1-, pubchem12871, sodium triacetoxy borohyride, sodium triacetyloxyboron 1-, tris acetoxy hydroborate sodium salt, sodium bis acetyloxy-$l^ 2-boranuidyl acetate
IUPAC Name sodium bis(acetyloxy)boranuidyl acetate
SMILES [Na+].CC(=O)O[BH-](OC(C)=O)OC(C)=O

Specifications

Melting Point ∼110°C (decomposition)
Quantity 5 g
UN Number UN3396
Beilstein 4047608
Sensitivity Moisture sensitive
Solubility Information Soluble in dimethyl sulfoxide,methanol,benzene,toluene,terahydrofuran,dioxane and methylene chloride.
Formula Weight 211.94
Percent Purity 95%
Chemical Name or Material Sodium triacetoxyborohydride
Hazard Category H260-H315-H318
Hazard Statement GHS H Statement
H261-H228-H318-H335-H315
In contact with water releases flammable gas.
Flammable solid.
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P223-P231+P232-P264b-P280-P302+P352-P305+P351+P338-P310-P332+P313-P335+P334-P362-P370+P378q-P501c
DOTInformation Hazard Class: 4.3; Packaging Group: II
EINECSNumber 000-000-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.