Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium tripolyphosphate, 85%, pure, Tech.

Catalog No. p-42507
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle

CAS: 7758-29-4 | Na5O10P3 | 367.85 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-29-4
Molecular Formula Na5O10P3
Molecular Weight (g/mol) 367.85
MDL Number MFCD00003514
InChI Key HWGNBUXHKFFFIH-UHFFFAOYSA-I
Synonym sodium tripolyphosphate, sodium triphosphate, pentasodium triphosphate, stpp, thermphos, armofos, polygon, tripoly, rhodiaphos lv, thermphos n
PubChem CID 24455
IUPAC Name pentasodium;[oxido(phosphonatooxy)phosphoryl] phosphate
SMILES [O-]P(=O)([O-])OP(=O)([O-])OP(=O)([O-])[O-].[Na+].[Na+].[Na+].[Na+].[Na+]

Specifications

Melting Point 622.0°C
Color White
Physical Form Crystalline Powder
Quantity 500 g
Assay Percent Range 85%
Linear Formula Na5P3O10
Merck Index 15, 8824
Solubility Information Solubility in water: 145g/L (20°C)
Formula Weight 367.85
Percent Purity 85%
Grade Pure
Packaging Plastic bottle
Chemical Name or Material Sodium tripolyphosphate, tech., 85%
Show More Show Less
Hazard Category HYGROSCOPIC
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 231-838-7
RTECSNumber YK4570000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.