Alfa Aesar™ Sodium tripolyphosphate

Manufacturer: Alfa Aesar™ 1344036

Quantity 500g

 View more versions of this product

Catalog No. AA1344036

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7758-29-4
    Molecular Formula Na5P3O10
    Formula Weight 367.91
    MDL Number MFCD00003514
    Merck Index 14,8697
    Synonym Sodium phosphate; Sodium triphosphate
    Chemical Name or Material Sodium tripolyphosphate

    Specifications

    Formula Weight 367.91
    Molecular Formula Na5P3O10
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Odor Odorless
    Synonym Sodium phosphate; Sodium triphosphate
    RTECS Number YK4570000
    Physical Form Granular
    Sensitivity Hygroscopic
    MDL Number MFCD00003514
    Color White
    Quantity 500g
    TSCA TSCA
    Solubility Information Soluble in water
    Health Hazard 1 Warning
    CAS 7758-29-4
    Merck Index 14,8697
    Melting Point 622°C
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material Sodium tripolyphosphate

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit