Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quidel Sofia 2 Flu+SARS Antigen FIA (25T)
SDP

Catalog No. 23044155 Shop All QuidelOrtho Products
Change view
Click to view available options
Quantity:
25
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-044-155 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-044-155 Supplier Quidel Supplier No. 20377
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Uses advanced immunofluorescence-based lateral flow technology in a sandwich design for qualitative detection of nucleocapsid protein from influenza A and influenza B, and SARS-CoV-2. Allows for testing of patients suspected of influenza A, influenza B, and COVID-19/2019-nCoV at the Point of Care.

  • One Sample - Three Results
  • Results in 15 minutes
  • Dual Work Modes
  • Virena Connection
  • All necessary components included in kit
  • Self-contained Test cassette
  • For use with Sofia 2
  • FDA Emergency Use Authorization.

Specifications

Certifications/Compliance End-User Placement Agreement Required
Content And Storage 15°C to 35°C
Control Sets Included and available for purchase
CE Marker No
Clia Complexity Waived (EUA)
Detectable Analytes Influenza A, Influenza B, COVID-19/2019-nCOV
Sample Type Direct: Nasal Swab, Nasopharyngael Swab
Sensitivity Influenza A: NS - 90%, NPS - 97.1%Influenza B: NS - 89%, NPS - 90%SARS-CoV-2: 95.2%
Type Rapid Test
For Use With (Application) Instrument Read - advanced immunofluorescence-based lateral flow
Packaging Type 12 Kits/Cs.
Quantity 25
Specificity Influenza A: NS - 95%, NPS - 94.6%Influenza B: NS - 96%, NPS - 97%SARS-CoV-2: 100%
Test Time 15 min.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.