Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quidel Sofia™ Strep A FIA
GSA_VASDP

Catalog No. p-4666348 Shop All QuidelOrtho Products
Click to view available options
Quantity:
25 Tests
Type:
Streptococcus A FIA
This item is not returnable. View return policy
This item is not returnable. View return policy

Includes

25 Test-Kit

  • Individually Packaged Cassettes (25): Polyclonal rabbit anti-Group A Streptococcus antibodies
  • Reagent Tubes (25)
  • Reagent Solution Bottles (25): 4M Sodium Nitrite and 0.2M Acetic Acid inside glass ampoule
  • Fixed Volume Pipettes (25)
  • Sterile Rayon Throat Swabs (25)
  • Positive Control Swab (1): Swab is coated with heat-inactivated, non-infectious Group A Streptococcus
  • Negative Control Swab (1): Swab is coated with heat-inactivated, non-infectious Group C Streptococcus
  • Package Insert (1)
  • Quick Reference Instructions (1)
  • QC Card (located on kit box)
  • Printer Paper (1)

For rapid qualitative detection of Group A Streptococcal antigens from throat swabs of symptomatic patients

The Sofia Strep A FIA employs immunofluorescence technology that is used with the Sofia Analyzer to detect Group A Streptococcal antigen.

  • For use with Sofia Analyzer
  • CLIA-Moderate
  • Highly sensitive and specific with enhanced assay accuracy
  • Analyzes and provides objective result versus visually read tests
  • Involves antigenic components extraction of Group A Streptococcus (GAS) bacteria
  • Results in 5 minutes

Applications:

Healthcare industry for diagnostic purposes

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Content And Storage Room Temperature
Detectable Analytes Streptococcal Antigen (Group A)
Sample Type Throat Swab
Type Streptococcus A FIA
Includes Cassettes, reagent tubes, reagent solution (contains Sodium Nitrite and Acetic Acid), reagent dropper tips, sterile rayon throat swabs, positive control swab and negative control swab
Quantity 25 Tests
Test Time 5 min.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.