Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ SoftFit-L™ Pipette Tips, TipCycle Recycle Kit, Hinged Racks

Click to view available options
Volume (Metric):
20 μL
200 μL
300 μL
1000 μL
1200 μL
Sterility:
Non-sterile
Sterile
Description
Gain the accuracy and precision you expect from your pipette samples every time with Thermo Scientific™ SoftFit-L™ pipette tips. For use with Rainin™ LTS pipettes.
- Recycle nonhazardous pipette tips, racks, and associated packaging with Tipcycle
- Complete TipCycle Pipette Tip Recycling Kit Includes: 1 case of pipette tips, box liner, liner tie, FedEx return shipping label, waiver form, and instructions for return
- Hinged rack provides easy one-hand access to tips
- Color-coded inserts allow quick identification of tip volume

Specifications
Specifications
Volume (Metric) | 1200 μL |
Filtered | Non-filtered |
Sterility | Non-sterile |
Color | Natural |
Autoclavable | Not Autoclavable |
Model | 1200 μL, Non-Filtered, Non-sterile, Hinged Rack, TipCycle Recycle Kit |
Length (English) | 3.42 in. |
Length (Metric) | 8.67 cm |
Clean Claims | DNase, RNase, DNA, Pryogen Free |
Format | Hinged Rack |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction