Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SOGA1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546498
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546498 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546498 Supplier Invitrogen™ Supplier No. PA546498
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: EPGDPEKDTK EKPGLSSRDC NHLGALACQD PPGRQMQRSY TAPDKTGIRV Sequence homology: Guinea Pig: 82%; Human: 100%; Rat: 77%.

SOGA1 regulates autophagy by playing a role in the reduction of glucose production in an adiponectin- and insulin-dependent manner.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SOGA1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Soga1
Gene Accession No. O94964
Gene Alias 80-kDa SOGA fragment; 9830001H06Rik; AI426038; C20orf117; C-terminal 80 kDa form; D430036N24Rik; dJ132F21.1; KIAA0889; N-terminal form; Protein SOGA1; Soga; SOGA family member 1; Soga1; Suppressor of glucose by autophagy; suppressor of glucose from autophagy; suppressor of glucose, autophagy associated 1; suppressor of glucose, autophagy-associated protein 1
Gene Symbols Soga1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human SOGA1 (aa 965-1014).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140710
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.