Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SOGA1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560576
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560576 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560576 Supplier Invitrogen™ Supplier No. PA560576
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LPGLREQAAL VSKAIDVLVA DANGFTAGLR LCLDNECADF RLHEAPDNSE GPRDTKLIHA ILVRLSVLQQ ELNAFTRKAD AVLGCSVKEQ QESF Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 89%.

SOGA1 regulates autophagy by playing a role in the reduction of glucose production in an adiponectin- and insulin-dependent manner.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SOGA1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Soga1
Gene Accession No. O94964
Gene Alias 80-kDa SOGA fragment; 9830001H06Rik; AI426038; C20orf117; C-terminal 80 kDa form; D430036N24Rik; dJ132F21.1; KIAA0889; N-terminal form; Protein SOGA1; Soga; SOGA family member 1; Soga1; Suppressor of glucose by autophagy; suppressor of glucose from autophagy; suppressor of glucose, autophagy associated 1; suppressor of glucose, autophagy-associated protein 1
Gene Symbols Soga1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SOGA1. Recombinant protein control fragment (Product #RP-99438).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140710
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.