Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SOGA1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA572776
Description
SOGA1 Polyclonal Antibody for Western Blot, ICC/IF, ELISA
SOGA1 regulates autophagy by playing a role in the reduction of glucose production in an adiponectin- and insulin-dependent manner.
Specifications
SOGA1 | |
Polyclonal | |
Unconjugated | |
Soga1 | |
80-kDa SOGA fragment; 9830001H06Rik; AI426038; C20orf117; C-terminal 80 kDa form; D430036N24Rik; dJ132F21.1; KIAA0889; N-terminal form; Protein SOGA1; Soga; SOGA family member 1; Soga1; Suppressor of glucose by autophagy; suppressor of glucose from autophagy; suppressor of glucose, autophagy associated 1; suppressor of glucose, autophagy-associated protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
140710 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
O94964 | |
Soga1 | |
SOGA1 antibody for a 16 amino acid peptide near the carboxy terminus of human SOGA. The immunogen is within the last 50 amino acids of SOGA1. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction