Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SOGA1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595810
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595810 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595810 Supplier Invitrogen™ Supplier No. PA595810
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AEVLPGLREQ AALVSKAIDV LVADANGFTA GLRLCLDNEC ADFRLHEAPD NSEGPRDTKL IHAILVRLSV LQQELNAFTR KADAVLGCSV KEQQESFSSL P; Positive Samples: Mouse liver, Mouse brain, Mouse lung, Rat liver, Rat brain.

SOGA1 regulates autophagy by playing a role in the reduction of glucose production in an adiponectin- and insulin-dependent manner.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SOGA1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.69 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Soga1
Gene Accession No. E1U8D0, O94964
Gene Alias 80-kDa SOGA fragment; 9830001H06Rik; AI426038; C20orf117; C-terminal 80 kDa form; D430036N24Rik; dJ132F21.1; KIAA0889; N-terminal form; Protein SOGA1; Soga; SOGA family member 1; Soga1; Suppressor of glucose by autophagy; suppressor of glucose from autophagy; suppressor of glucose, autophagy associated 1; suppressor of glucose, autophagy-associated protein 1
Gene Symbols Soga1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 650-750 of human SOGA1 (NP_5421942).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140710, 311578, 320706
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.