Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SOGA3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557254
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557254 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557254 Supplier Invitrogen™ Supplier No. PA557254
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GWRGKGVRAQ QRGGSGGEGA SPSPSSSSAG KTPGTGSRNS GSGVAGGGSG GGGSYWKEGC LQSELIQFHL KKER Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 81%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen SOGA3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SOGA3
Gene Accession No. Q5TF21
Gene Alias 6330407J23Rik; C6orf174; dJ403A15.3; Protein SOGA3; SOGA family member 3; SOGA3
Gene Symbols SOGA3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SOGA3. Recombinant protein control fragment (Product #RP-95037).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 387104
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.