Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SOGA3B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572219
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572219 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572219 Supplier Invitrogen™ Supplier No. PA572219
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SOGA3B Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen SOGA3B
Applications Western Blot
Classification Polyclonal
Concentration 1.58 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene soga3b
Gene Accession No. 0
Gene Alias fj45h03; protein SOGA3; uncharacterized protein LOC336052; uncharacterized protein soga3b; si:dkey-100j22.7; SOGA family member 3b; soga3b; wu:fj45h03; zgc:195224
Gene Symbols soga3b
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 198-229 amino acids from DANRE soga3b.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 336052
Target Species Zebrafish
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.