Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sohlh1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982097
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2097 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2097 Supplier Bioss Supplier No. BS12278R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Probable transcription factor required during spermatogenesis and oogenesis.

Specifications

Antigen Sohlh1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SOHLH1
Gene Accession No. Q5JUK2
Gene Alias bA100C15.3; bHLHe80; C9orf157; Gm110; helix-loop-helix protein NOHLH; newborn ovary helix loop helix; NOHLH; RP11-100C15.3; SOHLH1; SPATA27; spermatogenesis and oogenesis specific basic helix-loop-helix 1; Spermatogenesis- and oogenesis-specific basic helix-loop-helix-containing protein 1; spermatogenesis associated 27; TEB2; Tohlh1
Gene Symbols SOHLH1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Sohlh1 .
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 402381
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.