Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOHLH2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982098
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2098 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2098 Supplier Bioss Supplier No. BS12279R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Probable transcription factor, which may be involved in spermatogenesis and oogenesis.

Specifications

Antigen SOHLH2
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SOHLH2
Gene Accession No. Q9NX45
Gene Alias bHLHe81; SOHLH2; SPATA28; spermatogenesis and oogenesis specific basic helix-loop-helix 2; Spermatogenesis- and oogenesis-specific basic helix-loop-helix-containing protein 2; spermatogenesis associated 28; TEB1; testicular secretory protein Li 50; testicular secretory protein Li 51
Gene Symbols SOHLH2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human SOHLH2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54937
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.