Learn More
ICU Medical Sol-Care™ Safety Blood Collection Needle Infusion Set
Catalog No. p-8820013
Encompass
Click to view available options
Needle Gauge:
21 G
23 G
25 G
This item is not returnable. View return policy
This item is not returnable. View return policy
Safety butterfly needle designed to be used for blood collection.
  • Simple safety mechanism that should be activated immediately after blood withdrawal is complete
  • Reduces the risk of needlestick injury
  • Thumb rest to ease handling during safety activation
  • Flexible wings to secure device to patient's skin
  • Thin walled needle is designed to minimize patient discomfort
  • Available in 21, 23 or 25 gauge
  • Transparent housing and tubing for clear visualization of blood flow
  • DEHP-free, flexible, soft, and kink resistant tubing
Specifications
Product Type Needle Infusion Set
Color Green
Material PVC Tubing
Needle Gauge 21 G
Quantity 50 Pk., 4 Pk./Cs.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.