Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quidel Solana C. difficile AssayKit
GSA_VASDP

Catalog No. 23043121 Shop All QuidelOrtho Products
Change view
Click to view available options
Quantity:
48 Tests/Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-043-121 48 Tests/Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-043-121 Supplier Quidel Supplier No. M307
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

in vitro diagnostic test for the direct, qualitative detection of the Clostridium difficile Toxin A gene (tcdA) in unformed stool specimens

The Solana C. difficile Assay is intended for use as an aid in diagnosis of CDI. The assay utilizes helicase-dependent amplification (HDA) for the amplification of a highly-conserved fragment of the Toxin A gene sequence.

The Solana C. difficile Assay combines simple sample processing and Helicase-Dependent Amplification (HDA) performed in Solana for the detection of toxigenic Clostridium difficile directly from CDI-suspected diarrheal specimens.

In Solana, the target sequence is amplified by specific primers and detected by a specific fluorescence probe included in the Reaction Tube. A competitive process control (PRC) is included in the Lysis Tube to monitor sample processing, inhibitory substances in clinical samples, reagent failure or device failure. The PRC is amplified by the target-specific primers and detected by a PRC specific fluorescence probe.

Results are displayed on the touchscreen, can be saved to the instrument, printed, and are capable of being sent to the LIS and exported through one of Solana’s four USB ports. Solana C. difficile is supported by the power of Virena™

  •  
  • HDA technology
  • Lyophilized Master Mix reagent
  • Small, easy-to-use instrument
  • CLIA moderately complex
  • Refrigerated storage
  • Raw stool sample
  • LOD:
    • ATCC BAA-1805 9.13E+03 CFU/mL
    • CCUG 20309 4.90E+03CFU/mL
    • Genomic DNA: ATCC™ BAA-1382DQ™ 15 copy/assay

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Content And Storage 25°C ± 2° for up to 1 day or 2°C to 8°C for up to 6 days in sterile container
Sample Type Unformed Stool
Sensitivity 93%
Type C. difficile Assay
For Use With (Application) Direct, qualitative detection of the Clostridium difficile Toxin A gene (tcdA)
Quantity 48 Tests/Kit
Sample Size 2.0 mL
Specificity 0.992
Test Time 30 min.
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.