Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quidel Solana™ Flu Assay Startup Kit
SDP

Catalog No. p-6977026 Shop All QuidelOrtho Products
Change view
Click to view available options
Type:
Influenza A and B Assay Kit
Molecular Diagnostic Instrument
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
23-043-106 Molecular Diagnostic Instrument
23-043-107 Influenza A and B Assay Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-043-106 Supplier Quidel Supplier No. M917
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Includes

Instrument, Influenza A&B Test Kit, Heat Block, Printer, Labels, Tray, Racks & Stylus

For rapid differential diagnosis of acute influenza A and influenza B viral infections.

  • CLIA Complexity: Moderate, requires minnimal training
  • Small, ready to use benctop instrument
  •  LIS & middleware capable with Barcode Technology
  • Helicase-dependent Isothermal amplification (HDA) with fluorescence detection
  • For use with Solania&trade, Influenza A+B Assay Kit, Quidel Molecular Influenza A+B Control Set
  • For in vitro diagnostic use



Order Info

Order eligibility is based upon executed contractual agreement. Please reach out to your Fisher HealthCare Account Representative for more information.

Specifications

Content And Storage 2°C to 8°C
Control Sets M106
CPT Code 87502
Description null
CE Marker Yes
Clia Complexity Moderate
Detectable Analytes Influenza A and B
Sample Type Nasal Swab, Nasopharyngeal Swab
Sensitivity 98.6.%/100.0% (Culture), 97.2%/100% (PCR)
Type Molecular Diagnostic Instrument
For Use With (Application) Helicase-dependent amplification (HDA) with fluorescence detection
Includes Instrument, Influenza A&B Test Kit, Heat Block, Printer, Labels, Tray, Racks & Stylus
Quantity 1
Specificity 95.1.% / 99.3% (culture)  96.8%/98.9% (PCR)
Test Time ≈45 min.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.