Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quidel Solana RSV+hMPV Assay Kit
SDP

Catalog No. 23043123 Shop All QuidelOrtho Products
Change view
Click to view available options
Quantity:
48/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-043-123 48/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-043-123 Supplier Quidel Supplier No. M306
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Solana RSV + hMPV Assay is a rapid qualitative in vitro diagnostic test for the detection and differentiation of Respiratory Syncytial Virus (RSV) and Human Metapneumovirus (hMPV) viral RNA in nasal and nasopharyngeal swabs from patients with signs and symptoms of respiratory infection.

  • RT-HDA technology - Rapid method of isothermal nucleic acid amplification that does not require thermal cycler
  • Lyophilized Master Mix reagent - Easy-to-use format, just add processed sample to rehydrate
  • Small, easy-to-use instrument - Eliminates the need for dedicated molecular space and costly capital equipment, allowing for testing in smaller labs
  • CLIA moderately complex - Requires minimal training
  • Refrigerated storage - No freezer needed, 2°C to 8°C storage
  • Long shelf life - Currently 12 months from date of manufacture
  • Room temperature set up - No ice or cooling block required

Specifications

Content And Storage 2°C to 8°C
Sample Type Nasal swabs and nasopharyngeal swabs in transport media (BD/Copan VTM, Remel M4, Remel M4RT, Remel M
Includes Process Buffer 48 tubes/kit,Reaction Tubes 48 tubes/kit
Quantity 48/Pk.
Sensitivity Drift BD/Copan VTM, Remel M4, Remel M4RT, Remel M5, Remel M6, or Copen eSwab
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.