Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quidel Solana SARS-CoV-2 LYO Assay
SDP

Catalog No. p-7227706
Click to view available options
Includes:
48 Tests Only
48 Tests, 50 UTM/NS swab
48 Tests, 50 UTP/NP swab
Starter Kit- 48 Tests, 50 UTM Swab/ Analyzer
This item is not returnable. View return policy
This item is not returnable. View return policy

An isothermal Reverse Transcriptase - Helicase-Dependent Amplification (RT-HDA) assay intended for the qualitative detection of nucleic acid from SARS-CoV-2 in nasopharyngeal (NP) and nasal (NS) swab specimens from individuals suspected of COVID-19.

  • RT-HDA technology: Rapid method of isothermal nucleic acid amplification that does not require thermal cycler
  • Small, easy-to-use instrument
  • Room temperature set up
  • Long shelf life: 6 months from date of manufacture

Specifications

CE Marker CE-IVD
Clia Complexity Moderate
Detectable Analytes SARS-CoV-2
Format Tube
Disposable Single-use
For Use With (Application) RT-PCR, Rapid PCR
Includes 48 Tests Only
Sterility Non-sterile
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.