Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quidel Solana™ STREP Complete Kit
GSA_VASDP

Catalog No. p-6987018 Shop All QuidelOrtho Products
Click to view available options
Type:
Molecular Diagnostic Instrument
Strep Complete Assay
This item is not returnable. View return policy

Catalog No. 23-043-109

Supplier: Quidel M305

Only null left
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Includes

Test Kit: Package insert, Dilution Buffer, Lysis Buffer, Reaction Tubes

Features rapid method of nucleic acid amplification that does not require thermal cycler. For qualitative detection of Group A β-hemolytic Streptococcus (Streptococcus pyogenes), and pyogenic Group C/G (streptococcus dysgalactiae) nucleic acids.

  • Rapid in vitro diagnostic test for qualitative detection of Group A β-hemolytic Streptococcus (Streptococcus pyogenes), and pyogenic Group C/G (streptococcus dysgalactiae) nucleic acids
  • Isolated from throat swab specimens obtained from patients with signs and symptoms of pharyngitis, such as sore throat.
  • Throat swab sample or an aliquot of liquid transport media is lysed by simple heat treatment, diluted, and added to two Reaction Tubes, one containing primers and probes specific for Group A Strep
  • Results are displayed on the touchscreen,
  • Can be saved to the instrument, printed
  • Capable of being sent to the LIS and exported through one of Solana's four USB ports.
  • Solana Strep Complete is supported by the power of Virena™

Features and Benefits

  • Lyophilized Master Mix reagent: Easy-to-use format, just add processed sample to rehydrate
  • Small, easy-to-use instrument: Eliminates the need for dedicated molecular space and costly capital equipment, allowing for testing in smaller labs
  • CLIA moderately complex: Requires minimal training
  • Refrigerated storage: No freezer needed, 2°C to 8°C storage
  • Long shelf life: Currently 12 months from date of manufacture
  • Room temperature at set up: No ice or cooling block required
  • HDA Technology
  • No need for Culture Confirmation

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Certifications/Compliance CLIA-Moderate
Content And Storage 2°C to 8°C (No ice or cooling block required)
Clia Complexity Moderate
Detectable Analytes Streptococci (Group A, C, G)
Sample Type Throat Swab
Sensitivity 98.8% (Pyogenes), 100% (Dysgalactiae)
Type Strep Complete Assay
For Use With (Application) For diagnosis of Group A, C, & G Streptococcus
Includes Test Kit: Package insert, Dilution Buffer, Lysis Buffer, Reaction Tubes; Lyophilized Master Mix reagent
Quantity 48 Reactions/Pk.
Specificity 98.9% (Pyogenes)/99.5% (dysgalactiae)
Test Time ≈35 min.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.