Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Solanezumab Antibody (2372A) [Biotin], Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB10124B
Description
Solanezumab Monoclonal antibody specifically detects Solanezumab in Broad Species Reactivity samples. It is validated for Western Blot, Flow Cytometry, CyTOFSpecifications
Solanezumab | |
Monoclonal | |
Biotin | |
PBS | |
Full-length Solanezumab | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Flow Cytometry, CyTOF | |
2372A | |
Western Blot, Flow Cytometry, CyTOF-ready | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
Broad Species Reactivity | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction