Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Anti- Solanezumab (Anti-Idiotype) Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: R&D Systems MAB10124100
This item is not returnable.
View return policy
Description
Solanezumab Monoclonal specifically detects Solanezumab in Multi-Species samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
Solanezumab | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
Full-length Solanezumab | |
100 μg | |
Detects Solanezumab in Western blots and direct ELISAs. In Western blots, no cross-reactivity with Trastuzumab and Adalimumab is observed. | |
All species | |
Supernatant |
Western Blot, Flow Cytometry, CyTOF | |
2372A | |
Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction