Learn More
United Scientific™ Solid Glass Stoppers
SDPSCI_ED
Catalog No. S012414 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Standard Taper Joint:
#13
#16
#19
#22
#27
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Standard Taper Joint
S012414 #27
S012418 #13
S012417 #16
S012416 #19
S012415 #22
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. S012414 Supplier United Scientific™ Supplier No. UNSGS27
Only null left
Add to Cart
Add to Cart
Easily seal ground jointed glassware like flasks and separatory funnels with these Solid Glass Stoppers, featuring interchangeable ground joints.
  • Solid penny head glass stoppers for use with ground jointed glassware, such as flasks and separatory funnels
  • For ASTM standard flasks
  • Made of borosilicate 3.3 glass
Specifications
Standard Taper Joint #27
Material Borosilicate Glass
Shape Tapered
Stopper Number 27
Autoclavable Autoclavable
Length (Metric) 57 mm
Diameter (Metric) Outer 25 mm
Color Clear
Product Type Solid Pennyhead Stopper