Learn More
United Scientific™ Solid Glass Stoppers
SDPSCI_ED
Easily seal ground jointed glassware like flasks and separatory funnels with these Solid Glass Stoppers, featuring interchangeable ground joints.
Supplier:  United Scientific™ UNSGS16
 View more versions of this product
Catalog No. S012417
Only null left
Add to Cart
Description
Description
  • Solid penny head glass stoppers for use with ground jointed glassware, such as flasks and separatory funnels
  • For ASTM standard flasks
  • Made of borosilicate 3.3 glass
Specifications
Specifications
#16
Tapered
42 mm
15 mm
Solid Pennyhead Stopper
16
Clear
Autoclavable
Borosilicate Glass
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.