Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Supelco™ Solid Top Screw Cap with PTFE Liner
Shop All MilliporeSigma Supelco Products
Click to view available options
For Use With (Equipment):
2 mL Large Opening Vial
2 mL Standard Opening Vial
4 mL Vial
Description
- The color green easily identifies the PTFE liner system. F-217 foam-backed PTFE liners resist attack from virtually all chemicals at room temperature and provides odor and impact resilience for a tight seal with a low moisture transmission rate.
- F-217 has a low density foam core between two solid layers of LDPE (low density polyethylene).
- Some uses for these F-217/PTFE lined caps include analytical lab samples, high purity chemicals, strong acids solvents, environmental samples, pharmaceuticals, and diagnostic reagents.

Specifications
Specifications
Material (Liner) | F217/PTFE |
Color | Green |
Material | Melamine Resin |
No. per Pack | 100 |
Type | Solid Cap |
Closure Size | 8-425 |
For Use With (Equipment) | 2 mL Standard Opening Vial |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction