Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Solids Drinking Water/Wastewater Standard (ULTRAcheck™ Blind QC Standard)

Catalog No. usqci711
Encompass
Click to view available options
Quantity:
500 mL

Specifications

Chemical Name or Material Solids Drinking Water/Wastewater Standard
Name Note ULTRAcheck Blind QC Standard
Packaging Poly Bottle
Quantity 500 mL
Concentration or Composition (by Analyte or Components) Component concentrations in ready-to-use solution:
Suspended Solids    •20-200mg/L
Dissolved Solids    •50-5000mg/L
Total Solids    •70-5200mg/L
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.