Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAPA™ Solo Blue 999 Nitrile Gloves

Catalog No. 19046597
Click to view available options
Size:
Medium
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Powder free nitrile gloves are used in hospitals, food processing, and industrial applications where contamination is a concern.

  • Textured fingertips for improved dexterity
  • Easy to don on and take off
  • Latex-free
  • Previous name was Solo Ultra 999
  • Ideal for automotive/mechanics industry, aeronautics industry and laboratories

Specifications

Certifications/Compliance FDA 21CFR 177.2600
Certifications/Compliance (Europe) EN-ISO 374-5
Chemotherapeutic Rated No
Applications Healthcare, Laboratory, Industrial
Color Blue
Disposable Single-use
Exterior Finish Partially Textured
Latex Free Yes
Material Nitrile
Size Medium
Thickness (English) 4 mil
Thickness (Metric) 0.10 mm
FDA 510 (k) Yes
Model 34999437
Series Solo Blue 999
Length (English) 11.5 in.
Length (Metric) 29.2 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.