Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Soluble Adenylate Cyclase 1 Rabbit anti-Human, Mouse, Rat, Polyclonal, FabGennix

Catalog No. 502208945
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-8945 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-8945 Supplier Fabgennix Inc Supplier No. SAC121AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Soluble Adenylate Cyclase 1 Polyclonal antibody specifically detects Soluble Adenylate Cyclase 1 in Human, Mouse, Rat samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry, Immunomicroscopy, Immunoprecipitation, Western Blot

Specifications

Antigen Soluble Adenylate Cyclase 1
Applications ELISA, Immunocytochemistry, Immunohistochemistry, Immunomicroscopy, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene ADCY1
Gene Accession No. O88444, Q08828
Gene Alias 3',5'-cyclic AMP synthetase; Ac1; ADCY1; adenyl cyclase; adenylate cyclase 1; adenylate cyclase 1 (brain); adenylate cyclase type 1; Adenylate cyclase type I; adenylyl cyclase 1; ATP pyrophosphate-lyase 1; barrelless; brl; Ca(2+)/calmodulin-activated adenylyl cyclase; D11Bwg1392e; DFNB44; I-AC; mKIAA4070
Host Species Rabbit
Immunogen Synthetic peptide corresponding to unique amino acid sequence on SAC protein.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 107, 305509, 432530
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.