Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Soluble Guanylyl Cyclase alpha 2 Rabbit anti-Human, Mouse, Rat, Polyclonal, FabGennix

Catalog No. 502209078
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-9078 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-9078 Supplier Fabgennix Inc Supplier No. SGCA112AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Soluble Guanylyl Cyclase alpha 2 Polyclonal antibody specifically detects Soluble Guanylyl Cyclase alpha 2 in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot

Specifications

Antigen Soluble Guanylyl Cyclase alpha 2
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene GUCY1A2
Gene Accession No. P33402, Q9WVI4
Gene Alias 6330407I18Rik; A230060L24Rik; GC-SA2; GCS-alpha-2; guanylate cyclase 1 soluble subunit alpha 2; guanylate cyclase 1, soluble, alpha 2; guanylate cyclase soluble subunit alpha-2; GUC1A2; GUCSA2; Gucy1a2; soluble guanylyl cyclase alpha2 subunit
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 680-730 on rat guanylate cyclase 1, soluble, alpha 2 protein.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 234889, 2977, 66012
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.