Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Solution dNTPs: PCR Nucleotide Mix

Free from DNase, RNase, and nicking enzyme activity

$162.80 - $303.05

Specifications

Format Liquid
Purity >99%
Color Colorless
For Use With (Application) Functionally tested for long PCR and sequencing, Nucleic Acid amplification, labelling, mutagenesis, cDNA synthesis and expression profiling
View More Specs

Products 2
Catalog Number Mfr. No. Concentration Product Type Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Concentration Product Type Quantity Price Quantity & Availability  
45-001-459
View Documents
Cytiva
28406560
25 mM PCR Nucleotide Mix dNTP Mix (25mM each A,C,G,T) 500 μm
Each for $303.05
Only null left
Add to Cart
 
45-001-461
View Documents
Cytiva
28406564
10 mM PCR Nucleotide Mix dNTP Mix (10mM each A,C,G,T) 500 μL
Each for $162.80
Only null left
Add to Cart
 
Description

Description

  • Greater than 99% triphosphate purity (by HPLC) for optimum performance and consistency
  • Buffer-free and ready-to-use solutions in multiple formats
  • Functionally tested for long PCR and sequencing

Order Info

Custom formulation, testing, packaging, and labeling are available

Specifications

Specifications

Liquid
>99%
Colorless
Functionally tested for long PCR and sequencing, Nucleic Acid amplification, labelling, mutagenesis, cDNA synthesis and expression profiling
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.