Learn More
ENG Scientific Solution II - Acid Alcohol (3% HCL)
Solution II - Acid Alcohol (3% HCL) is ideal for staining acid-fast bacteria. Use with Acid-fast stain kit. Modification of the Kinyoun Method.
Supplier: ENG Scientific 6512
This item is not returnable.
View return policy
Description
Specifications
| Bottles | |
| 1 gal. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For in vitro diagnostic use only.
Spot an opportunity for improvement?Share a Content Correction