Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Solvent blue 35, MP Biomedicals™

Catalog No. icn15663550
Click to view available options
Quantity:
50 g
100 g

Chemical Identifiers

CAS 17354-14-2
Molecular Formula C22H26N2O2
Molecular Weight (g/mol) 350.462
MDL Number MFCD00011714
InChI Key OCQDPIXQTSYZJL-UHFFFAOYSA-N
Synonym C.I. 61554, Sudan Blue II
PubChem CID 3766139
IUPAC Name 1,4-bis(butylamino)anthracene-9,10-dione
SMILES CCCCNC1=C2C(=C(C=C1)NCCCC)C(=O)C3=CC=CC=C3C2=O

Specifications

Formula Weight 350.5
Density 1.23g/mL
Quantity 50 g
Physical Form Powder
Chemical Name or Material Solvent Blue 35, Dye content: Approx. 90%
EINECSNumber 241-379-4
Recommended Storage Store at Room Temperature (15-30°C).
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.