Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Solvent Yellow 124, ≥98.0% (HPLC), MilliporeSigma™ Supelco™
Click to view available options
Quantity:
50 mg
250 mg
Description
Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications
Specifications
Packaging | Bottomless Glass Bottle with Inserted Fused Cone |
CAS | 34432-92-3 |
Physical Form | Neat |
Linear Formula | C22H31N3O2 |
Synonym | N-Ethyl-N-[2-(1-isobutoxyethoxy)ethyl]-p-(phenylazo)aniline |
Type | Analytical Standard |
Quantity | 50 mg |
Formula Weight | 369.50 |
Percent Purity | ≥98.0% (HPLC) |
Grade | Analytical Standard |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction