Learn More
Somatostatin Monoclonal Antibody (ICDCLS), eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$158.65 - $326.25
Specifications
Antigen | Somatostatin |
---|---|
Clone | ICDCLS |
Concentration | 0.5 mg/mL |
Applications | Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-112-9323
![]() |
Invitrogen™
14975180 |
25 μg |
Each
for $158.65
Save 9% Reg : |
|
|||||
50-112-9262
![]() |
Invitrogen™
14975182 |
100 μg |
Each
for $326.25
Save 25% Reg : |
![]() |
|||||
Description
Description: The monoclonal antibody ICDCLS recognizes human, mouse, and rat Somatostatin. Somatostatin, also known as growth hormone-inhibiting hormone (GHIH) or somatotrophin release-inhibiting factor (SRIF), is a peptide hormone with two active forms (14 and 28 amino acids) produced by alternative cleavage of a single preproprotein. Somatostatin is secreted by the delta cells of the islet of Langerhans in the pancreas as well as by the hypothalamus and intestine. Somatostatin has a variety of functions, such as regulating the endocrine system and affecting neurotransmission and cell proliferation. Somatostatin also inhibits the release of numerous secondary hormones, such as insulin and glucagon. Somatostatin can be used to identify medium size aspiny neurons in the striatum, cortex, and hippocampus. Expression of somatostatin is present in hyperplasia of the pancreatic islets as well as islet cell tumors such as somatostatinomas. Applications Reported: This ICDCLS antibody has been reported for use in immunohistochemical staining of formalin-fixed paraffin embedded tissue sections, and microscopy. Applications Tested: This ICDCLS antibody has been tested by immunohistochemistry of formalin-fixed paraffin embedded human tissue using low pH antigen retrieval and can be used at less than or equal to 0.1 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
The hormone somatostatin has active 14 aa and 28 aa forms that are produced by alternate cleavage of the single preproprotein encoded by this gene. Somatostatin is expressed throughout the body and inhibits the release of numerous secondary hormones by binding to high-affinity G-protein-coupled somatostatin receptors. This hormone is an important regulator of the endocrine system through its interactions with pituitary growth hormone, thyroid stimulating hormone, and most hormones of the gastrointestinal tract. Somatostatin also affects rates of neurotransmission in the central nervous system and proliferation of both normal and tumorigenic cells.Specifications
Somatostatin | |
0.5 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
Antrin; Growth hormone release-inhibiting factor; Neuronostatin; NST; preprosomatostatin; prepro-somatostatin; SMST; SOM; Somatostatin; somatostatin 28; somatostatin-14; Somatostatin-28; somatotropin release-inhibiting factor; SRIF; SS; SS-14; SS-28; Sst; SST-14 | |
SST | |
Primary | |
4°C | |
SST |
ICDCLS | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat | |
P60041, P60042, P61278 | |
20604, 24797, 6750 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.