Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Somatostatin Mouse anti-Human, Alexa Fluor 488, Clone: U24-354, BD Pharmingen™

Catalog No. BDB566032 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB566032 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB566032 Supplier BD Biosciences Supplier No. 566032
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Somatostatin Monoclonal antibody specifically detects Somatostatin in Human, Mouse, Rat samples. It is validated for Flow cytometry (Intracellular Staining)) .
TRUSTED_SUSTAINABILITY

Specifications

Antigen Somatostatin
Applications Flow Cytometry, Flow Cytometry (Intracellular Staining)
Classification Monoclonal
Clone U24-354
Concentration 0.2mg/mL
Conjugate Alexa Fluor 488
Formulation Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide.
Gene Alias growth hormone release-inhibiting factor, SST
Host Species Mouse
Immunogen Human Preprosomatostatin Recombinant Protein
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to Alexa Fluor™ 488 under optimum conditions, and unreacted Alexa Fluor™ 488 was removed.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Reseach Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.