Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Somatostatin Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA597200
Description
Immunogen sequence: TGAPSDPRLR QFLQKSLAAA AGKQELAKYF LAELLSEPNQ TENDALEPED LSQAAEQDEM RLELQRSANS NPAMAPRERK AGCKNFFWKT FTSC Positive Samples: HeLa, SW620, MCF7, Mouse intestine, Rat stomach.
The hormone somatostatin has active 14 aa and 28 aa forms that are produced by alternate cleavage of the single preproprotein encoded by this gene. Somatostatin is expressed throughout the body and inhibits the release of numerous secondary hormones by binding to high-affinity G-protein-coupled somatostatin receptors. This hormone is an important regulator of the endocrine system through its interactions with pituitary growth hormone, thyroid stimulating hormone, and most hormones of the gastrointestinal tract. Somatostatin also affects rates of neurotransmission in the central nervous system and proliferation of both normal and tumorigenic cells.
Specifications
Somatostatin | |
Polyclonal | |
Unconjugated | |
SST | |
Antrin; Growth hormone release-inhibiting factor; Neuronostatin; NST; preprosomatostatin; prepro-somatostatin; SMST; SOM; Somatostatin; somatostatin 28; somatostatin-14; Somatostatin-28; somatotropin release-inhibiting factor; SRIF; SS; SS-14; SS-28; Sst; SST-14 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
20604, 24797, 6750 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
2.47 mg/mL | |
PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 | |
P60041, P60042, P61278 | |
SST | |
Recombinant fusion protein containing a sequence corresponding to amino acids 23-116 of human SST (NP_0010391). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction